1,2,4-Trichlorobenzene ≥99% for HPLC, for GPC (gel permeation chromatography), J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

9444-05
JT9444-5EA 588.62 USD
JT9444-5
1,2,4-Trichlorobenzene ≥99% for HPLC, for GPC (gel permeation chromatography), J.T.Baker®
1,2,4-Trichlorobenzene

For high temperature gel permeation chromatography. UV cutoff at 310nm.

Formula: C₆H₃Cl₃
MW: 181.45 g/mol
Boiling Pt: 214.4 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1.45 g/cm³ (20 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00000547
CAS Number: 120-82-1
EINECS: 204-428-0
UN: 2321
ADR: 6.1,III
Merck Index: 13,09704

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₆H₃Cl₃) (by GC) ≥99 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤0.15
Ultraviolet Absorbance (1.00-cm cell vs. water) - 375 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm ≤0.010
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤310
Residue after Evaporation ≤5 ppm
Water (H₂O)(by Karl Fischer titrn) ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR