Tetrahydrofuran ≥99.5% (by GC, corrected for water content) stabilized for HPLC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 75 - 400 ppm

BAKR9440-03
JT9440-3CS 1676.71 USD
JT9440-3 JT9440-88
Tetrahydrofuran ≥99.5% (by GC, corrected for water content) stabilized for HPLC, J.T.Baker®
Tetrahydrofuran

For use in gel permeation chromatography. Stabilized with BHT (butylated hydroxytoluene; 2,6-di-tert-butyl-p-cresol). Packaged under nitrogen. Lot analysis on label.

Formula: C₄H₈O
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108.5 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (by GC, corrected for water, exclusive of preservative) ≥99.5 %
Color (APHA) ≤10
Peroxide (as H₂O₂) ≤0.005 %
Stabilizer (BHT) 75 - 400 ppm
Water (by KF, coulometric) ≤0.010 %
Stabilized with approximately 0.01% Butylated Hydroxytoluene


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR