Sodium hypochlorite 5% available chlorine in aqueous solution, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

9416-01 9416-03
JT9416-1CS 1068.75 USD
JT9416-1 JT9416-3
Sodium hypochlorite 5% available chlorine in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Sodium hypochlorite
Formula: ClNaO
Storage Temperature: Refrigerator
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance (Clear, light yellow liquid) Passes Test
Available Chlorine (when packaged) 5.0 - 6.0 %
Phosphate (PO₄) ≤0.0005 %
Calcium (Ca) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR