(±)-1,2-Propanediol ≥99.5% USP, Multi-Compendial

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane

Management of Change (MOC) category = R

9403-07 9403-03 9403-24 9403-26
JT9403-7EA 1696.3 USD
JT9403-7 JT9403-3 JT9403-24 JT9403-26
(±)-1,2-Propanediol ≥99.5% USP, Multi-Compendial
Propylene glycol
Formula: CH₃CH(OH)CH₂OH
MW: 76.1 g/mol
Boiling Pt: 187.6
Melting Pt: -59
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 99
Storage Temperature: Ambient
MDL Number: MFCD00064272
CAS Number: 57-55-6
EINECS: 200-338-0
Merck Index: 13,07947
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets U.S.P Requirements
Meets J.P. Chemical Specifications
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Specific Gravity at 25°/25°C 1.035 - 1.037
USP - Acidity Passes Test
USP - Water (H2O)(by Karl Fischer titrn) ≤ 0.20 %
USP - Identification A (IR) Passes Test
USP - Identification B - Diethylene Glycol ≤ 0.10 %
USP - Identification B - Ethylene Glycol ≤ 0.10 %
USP - Identification C Passes Test
USP - Residue on Ignition ≤ 0.007 %
USP - Chloride (Cl) ≤ 0.007 %
USP - Sulfate (SO₄) ≤ 0.006 %
USP - Assay (CH₃CHOHCH₂OH) ≥ 99.5 %
Endotoxin Concentration (EU/g) ≤ 2.5
EP/BP - Relative Density (d²º₂₀) 1.035 - 1.040
EP/BP - Refractive Index at 20°C 1.431 - 1.433
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Identification D Passes Test
EP/BP - Appearance Passes Test
EP/BP - Acidity Passes Test
EP/BP - Oxidizing Substances Passes Test
EP/BP - Reducing Substances Passes Test
EP/BP - Water (H₂O) ≤ 0.2 %
EP/BP - Ash (sulfated) ≤ 0.01 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Specific Gravity at 20°/20°C 1.035 - 1.040
JP - Acidity Passes Test
JP - Chloride (Cl) ≤ 0.007 %
JP - Sulfate (SO₄) ≤ 0.002 %
JP - Heavy Metals (as Pb) ≤ 5 ppm
JP - Arsenic (As) ≤ 2 ppm
JP - Glycerin Passes Test
JP - Water (H₂O) ≤ 0.5 %
JP - Residue on Ignition ≤ 0.005 %
JP - Distilling Range (Lower) 184 - 189 °C
JP - Distilling Range (Upper) 184 - 189 °C
JP - Ethylene Glycol ≤ 0.1 %
JP - Diethylene Glycol ≤ 0.1 %
JP - Any Other Impurity ≤ 0.1 %
JP - Total Impurities ≤ 1.0 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR