Tetrachloroethylene ≥99.8%, ULTRA RESI-ANALYZED® for organic trace analysis, for IR spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = N
9360-03
JT9360-3CS
2463.81
USD
JT9360-3
Tetrachloroethylene ≥99.8%, ULTRA RESI-ANALYZED® for organic trace analysis, for IR spectrophotometry, J.T.Baker®
Tetrachloroethylene
Formula:
C₂Cl₄ Boiling Pt: 121.1 °C (1013 hPa) Melting Pt: –22 °C Density: 1.63 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000834 CAS Number: 127-18-4 EINECS: 204-825-9 UN: 1897 ADR: 6.1,III Merck Index: 12,09332 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (Cl₂C:CCl₂) (by GC) | ≥99.8 % |
Infrared Absorbance Test: Hydrocarbon (exclusive of stabilizer) | ≤5 ppm |
Residue after Evaporation | ≤10 ppm |
Chloride (Cl) | ≤1.0 ppm |
Titrable Acid (µeq/g) | ≤0.5 |
Water (H₂O)(by Karl Fischer titrn) | ≤0.005 % |
Free Halogen (ASTM-D 4755) | Passes Test |
Range 3200-2700 cm⁻¹ | |
Measured at 2930 cm⁻¹ | |
(Hexadecane:isooctane:chlorobenzene standard mixture) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...