Toluene ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Methylbenzene, Methacide, Benzene, methyl, Phenylmethane, Methane, phenyl-, Toluol, FM approval, Methylbenzol
Management of Change (MOC) category = N
9351-02
9351-03
9351-33
JT9351-2CS
740.92
USD
JT9351-2
JT9351-3
JT9351-33
Toluene ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Toluene
Packaged under nitrogen. For use in high performance liquid chromatography. UV cutoff 285nm. Lot analysis on label.
- UV cutoff 285 nm
Formula:
C₆H₅CH₃ MW: 92.14 g/mol Boiling Pt: 110.6 Melting Pt: -95 Density: 0.867 g/cm³ (20 °C) Flash Pt: 4 Storage Temperature: Ambient |
MDL Number:
MFCD00008512 CAS Number: 108-88-3 EINECS: 203-625-9 UN: 1294 ADR: 3 II REACH: 05-2114615130-69 Merck Index: 13,09607 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (by GC, corrected for water) | ≥99.7 % |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm | ≤0.01 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 nm | ≤0.10 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 288 nm | ≤0.40 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm | ≤285 |
Residue after Evaporation | ≤3.0 ppm |
Substances Darkened by H₂SO₄ | Passes Test |
Water (by KF, coulometric) | ≤0.02 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...