Dichloromethane ≥99.5% (by GC, corrected for water content), Baker Analyzed™ ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

9324-22 9324-33 9324-05 9324-R 9324-03 9324-07 BAKR9324-01
JT9324-22CS 1888.47 USD
JT9324-22 JT9324-33 JT9324-5 JT9324-R JT9324-3 JT9324-7 SCJT9324-3 JT9324-01
Dichloromethane ≥99.5% (by GC, corrected for water content), Baker Analyzed™ ACS, J.T.Baker®
Dichloromethane

Lot analysis on label.

Formula: CH₂Cl₂
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CH₂Cl₂) (by GC, corrected for water) ≥99.5 %
Color (APHA) ≤10
Residue after Evaporation ≤0.0020 %
Titrable Acid (µeq/g) ≤0.3
Free Halogens Passes Test
Water (by KF, coulometric) ≤0.02 %
Product Information (not specifications):
Appearance (Clear, colorless liquid)
Boiling Point (typical)................. 39.5 °C.
Density (g/mL) at 25 °C (typical)........ 1.318
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR