4-Methyl-2-pentanone ≥98.5% (by GC, corrected for water content), BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

9322-01 9322-03
JT9322-1CS 1606.18 USD
JT9322-1 JT9322-3
4-Methyl-2-pentanone ≥98.5% (by GC, corrected for water content), BAKER ANALYZED® ACS
4-Methyl-2-pentanone

Lot analysis on label.

Formula: (CH₃)₂CHCH₂COCH₃
Boiling Pt: 117.4 °C (1013 hPa)
Melting Pt: –84 °C
Density: 0.801 g/cm³ (20 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00008938
CAS Number: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3,II
Merck Index: 13,05227
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d44947ef43cbc8103ae523ce5da380ef184ba09&vid=373b9075524004b95d32d3cf9698896714467e8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CH₃COCH₂CH(CH₃)₂) (by GC, corrected for water) ≥98.5 %
Color (APHA) ≤15
Residue after Evaporation ≤0.0050 %
Titrable Acid (meq/g) ≤0.002
Water (H₂O)(by Karl Fischer titrn) ≤0.1 %
Product Information (not specifications):
Appearance (Clear, colorless liquid)
Boiling Point (typical)................. 115.5 °C.
Density (g/mL) at 25 °C (typical)........ 0.796
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR