Dichloromethane ≥99.8% (by GC, corrected for water content) stabilised, BAKER ANALYZED® HPLC for HPLC, for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Stabilization: Stabilised with cyclohexene ≥ 50 ppm
Management of Change (MOC) category = N

9315-03 9315-33 9315-02
JT9315-3CS 1210.48 USD
JT9315-3 JT9315-33 JT9315-2
Dichloromethane ≥99.8% (by GC, corrected for water content) stabilised, BAKER ANALYZED® HPLC for HPLC, for spectrophotometry, J.T.Baker®
Dichloromethane

Methylene Chloride HPLC For use in liquid chromatography (HPLC and UHPLC) and spectrophotometry.

Formula: CH₂Cl₂
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm ≤0.01
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission For Information Only
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission For Information Only
Fluorescence Trace Impurities measured as Quinine Base at Emission Maximum for Impurities, For Information Only
Maximum for Impurities, For Information Only
Assay (by GC, corrected for water, exclusive of preservative) ≥99.8 %
Chloride (Cl) ≤10 ppm
Preservative (cyclohexene) ≥50 ppm
Residue after Evaporation ≤2.0 ppm
Titrable Acid (µeq/g) ≤0.3
Water (by KF, coulometric) ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR