Hexane (mixture of isomers) ≥98.5% (by GC, sum of enantiomers), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

9309-22 9309-05 9309-R 9309-33 9309-03 9309-07 9309-6Q 9309-ZX BAKR9309-01
JT9309-22CS 941.24 USD
JT9309-22 JT9309-5 JT9309-R JT9309-33 JT9309-3 JT9309-7 JT9309-6Q JT9309-ZX JT9309-01
Hexane (mixture of isomers) ≥98.5% (by GC, sum of enantiomers), BAKER ANALYZED® ACS, J.T.Baker®
Hexane (mixture of isomers)

Lot analysis on label.

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 68…70 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.663 g/cm³ (25 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Sum of 5 isomers, total hexanes plus methylcyclopentane) (by GC) ≥98.5 %
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.001 %
ACS - Water-Soluble Titrable Acid, meq/g ≤0.0003
ACS - Sulfur Compounds (as S) ≤0.005 %
ACS - Thiophene Passes Test
Product Information (not specifications):
Boiling Point (typical)................. 68.7 °C.
Density (g/mL) at 25 °C (typical)........ 0.663


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR