Cyclohexane ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Hexahydro-Benzene, Hexahydrobenzene, Hexamethylene

Management of Change (MOC) category = N

9292-03
JT9292-3CS 1728.86 USD
JT9292-3
Cyclohexane ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Cyclohexane

Packaged under nitrogen. Lot analysis on label. For high performance liquid chromatography. UV cutoff 205nm.


  • UV cutoff 205nm

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 81 °C (1013 hPa)
Melting Pt: 7 °C
Density: 0.7785 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm ≤0.08
Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm ≤0.20
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.30
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤205
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.5 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
Assay (C₆H₁₂) (by GC, corrected for water) ≥99.5 %
Residue after Evaporation ≤2.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR