ALCH, Proprietary Solvent III-1, anhydrous, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

9287-03 9287-33 9287-07
JT9287-3CS 1016.19 USD
JT9287-3 JT9287-33 JT9287-7
ALCH, Proprietary Solvent III-1, anhydrous, BAKER ANALYZED®, J.T.Baker®
Alcohol

Denatured with 1% ethyl acetate, 1% methyl isobutyl ketone, 1% gasoline, and 4 to 6% methanol. Lot analysis on label.


  • Total alcohol: 99.5%
  • Ethyl alcohol: 96%
  • Denaturant: methanol and a small amount of denatonium benzoate


SDA Formula 1-1 consists of 4 volumes of methanol, 1/8 oz of denatonium benzoate, and 100 volumes of 200 proof ethanol. * Specially Denatured Alcohol Formula 1-1 (200 Proof)(v/v)(parts by vol.)........ 100 *
Ethyl Acetate (v/v)(parts by vol)....... 0.87 * Methyl Isobutyl Ketone (v/v)(parts by vol)..... 1 * Rubber Hydrocarbon Solvent (v/v)(parts by vol)... 1

Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Color (APHA) ≤15
Residue after Evaporation ≤0.003 %
Specific Gravity at 60°/60°F 0.717 - 0.875
Water (H₂O)(by Karl Fischer titrn) ≤0.3 %
SDA Formula 1-1 consists of 4 volumes of methanol, 1/8 oz of denatonium
benzoate, and 100 volumes of 200 proof ethanol. * Specially Denatured
Alcohol Formula 1-1 (200 Proof)(v/v)(parts by vol.)........ 100 *
Ethyl Acetate (v/v)(parts by vol)....... 0.87 * Methyl Isobutyl Ketone
(v/v)(parts by vol)..... 1 * Rubber Hydrocarbon Solvent (v/v)(parts by
vol)... 1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR