Petroleum spirit, 20…40 °C, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

Management of Change (MOC) category = N

9272-03
JT9272-3CS 1848.78 USD
JT9272-3
Petroleum spirit, 20…40 °C, BAKER ANALYZED®, J.T.Baker®
Petroleum spirit
Boiling Pt: 20… 40 °C (1013 hPa)
Density: 0.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance and Color Passes Test
Residue after Evaporation ≤0.0020 %
Acidity Passes Test
Heavy Oils and Fats Passes Test
Boiling Range(Lower) 20 - 40 °C
Boiling Range(Upper) 20 - 40 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR