Dichloromethane ≥99.8% (by GC, corrected for water content) stabilized, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Methane dichloride, Methylene chloride, Methylene chloride

Stabilization: Stabilised with a methanol-alkene preservative system
Management of Change (MOC) category = NI

9264-02 9264-03
JT9264-2CS 1018.9 USD
JT9264-2 JT9264-3 JT9264-H1
Dichloromethane ≥99.8% (by GC, corrected for water content) stabilized, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Dichloromethane

For organic residue analysis. Stabilized with 400 to 600ppm methanol.

Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤5
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
Assay (CH₂Cl₂) (by GC, exclusive of preservative, corrected for water) ≥99.8 %
Color (APHA) ≤10
Residue after Evaporation ≤1.0 ppm
Titrable Acid (µeq/g) ≤0.3
Chloride (Cl) ≤10 ppm
Water (by KF, coulometric) ≤0.02 %
Contains a methanol-alkene preservative system.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR