Hexane (mixture of isomers) ≥95% (by GC, corrected for water content) (n-hexane ≥95%), BAKER ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Packaged under inert gas.

9262-02 9262-03 9262-3P 9262-7K 9262-8P 9262-8R 9262-92 9262-95 9262-98 9262-9A 9262-9V 9262-A3 9262-B5 9262-H1 9262-Q3 9262-S5 9262-V8 9262-YQ 9262-YA
JT9262-2CS 1018.16 USD
JT9262-2 JT9262-3 JT9262-3P JT9262-7K JT9262-8P JT9262-8R JT9262-92 JT9262-95 JT9262-98 JT9262-9A JT9262-9V JT9262-A3 JT9262-B5 JT9262-H1 JT9262-Q3 JT9262-S5 JT9262-V8 JT9262-YQ JT9262-YA
Hexane (mixture of isomers) ≥95% (by GC, corrected for water content) (n-hexane ≥95%), BAKER ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Hexane (mixture of isomers)

For organic residue analysis.

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –94.3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –23 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤5
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
ECD-Sensitive Impurities (as Ethylene Dibromide) - Single Impurity Peak (ng/mL) ≤5
Assay (Total Saturated C₆ Isomers) (by GC, corrected for water) ≥99.5 %
Assay (as n-Hexane) (by GC, corrected for water) ≥95 %
Color (APHA) ≤10
Residue after Evaporation ≤1.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤0.05 %
MEETS SPECIFICATIONS WITHIN THE EXPIRATION PERIOD


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR