Cyclohexane ≥99.0% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = NI

9258-03
JT9258-3CS 2064.4 USD
JT9258-3
Cyclohexane ≥99.0% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Cyclohexane

For organic residue analysis.

Formula: C₆H₁₂
Boiling Pt: 81 °C (1013 hPa)
Melting Pt: 7 °C
Density: 0.7785 g/cm³ (20 °C)
Flash Pt: –18 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Trace Organic Residues - FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤10
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
Assay (C₆H₁₂) (by GC, corrected for water) ≥99.0 %
Color (APHA) ≤10
Residue after Evaporation ≤2.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR