Diisopropyl ether ≥99.0% (by GC, corrected for water content) stabilized, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Isopropyl ether

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 0.01 %
Management of Change (MOC) category = N

9243-03 9243-22
JT9243-3CS 2631.87 USD
JT9243-3 JT9243-22
Diisopropyl ether ≥99.0% (by GC, corrected for water content) stabilized, BAKER ANALYZED® ACS, J.T.Baker®
Diisopropyl ether

Stabilized with approximately 0.01% hydroquinone. Lot analysis on label.

Formula: (CH₃)₂CHOCH(CH₃)₂
MW: 102.18 g/mol
Boiling Pt: 68.5 °C (1013 hPa)
Melting Pt: –86 °C
Density: 0.726 g/cm³ (20 °C)
Flash Pt: –28 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008880
CAS Number: 108-20-3
EINECS: 203-560-6
UN: 1159
ADR: 3,II
Merck Index: 13,05232
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=932b8c021b2bfa9a11b8d8812cafbafecc6f9239&vid=dea267e876203eb2c2674787701ec602da1b3cf6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay ((CH₃)₂CHOCH(CH₃)₂) (by GC, corrected for water) ≥99.0 %
Color (APHA) ≤25
Residue after Evaporation ≤0.0100 %
Titrable Acid (meq/g) ≤0.0007
Peroxide (as C₆H₁₄O₂) ≤0.05 %
Stabilized with approximately 0.01% Butylated Hydroxytoluene

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR