Diethanolamine ≥98.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

9227-01
JT9227-1CS 3551 USD
JT9227-1 JT9227-3
Diethanolamine ≥98.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Diethanolamine

Lot analysis on label.

Formula: C₄H₁₁NO₂
Boiling Pt: 268.8 °C (1013 hPa)
Melting Pt: 28 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD00002843
CAS Number: 111-42-2
EINECS: 203-868-0
REACH: 01-2119488930-28
Merck Index: 13,03134

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay ((HOCH₂CH₂)₂NH) (by GC, corrected for water) ≥98.5 %
ACS - Apparent Equivalent Weight 104.0 - 106.0
ACS - Color (APHA) ≤15
ACS - Monoethanolamine ≤1.0 %
ACS - Triethanolamine ≤1.0 %
ACS - Residue after Ignition ≤0.005 %
ACS - Heavy Metals (as Pb) ≤0.002 %
ACS - Iron (Fe) ≤0.001 %
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.15 %
Chloride (Cl) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR