Methyl ethyl ketone ≥99.5% (by GC), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: EK 500 Additive, Butan-2-one, Ethyl methyl ketone, 2-Butanone, Butanone, MEK

Management of Change (MOC) category = N

9214-03
JT9214-3CS 1078.4 USD
JT9214-3
Methyl ethyl ketone ≥99.5% (by GC), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Methyl ethyl ketone

Packaged under nitrogen. Lot analysis on label. For high performance liquid chromatography. UV cutoff 330nm.


  • UV cutoff 330 nm

Formula: C₂H₅COCH₃
Boiling Pt: 79.6 °C (1013 hPa)
Melting Pt: –86 °C
Density: 0.8054 g/cm³ (20 °C)
Flash Pt: –4 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3,II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-350 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 nm ≤0.07
Ultraviolet Absorbance (1.00-cm cell vs. water) - 335 nm ≤0.40
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤330
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.5 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
Assay (CH₃COCH₂CH₃) (by GC) ≥99.5 %
Residue after Evaporation ≤3.0 ppm
Water (by KF, volumetric) ≤0.03 %
Product Information (not specifications):
Windows of Infrared Transmittance (0.1-mm path,50-100% T) µm:
2.0-2.8; 2.9-3.2; 3.6-5.5; 9.4-9.8; 11.0-12.8; 13.5-15.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR