1-Butanol ≥99.4%, PHOTREX® for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Management of Change (MOC) category = N
9189-01
JT9189-1CS
1904.71
USD
JT9189-1
1-Butanol ≥99.4%, PHOTREX® for spectrophotometry, J.T.Baker®
1-Butanol
For spectrophotometry. Lot analysis on label.
Formula:
H₃C(CH₂)₂CH₂OH Boiling Pt: 117.6 °C (1013 hPa) Melting Pt: –89 °C Density: 0.812 g/cm³ (20 °C) Flash Pt: 30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002964 CAS Number: 71-36-3 EINECS: 200-751-6 UN: 1120 ADR: 3,III REACH: 01-2119484630-38 Merck Index: 13,01539 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (CH₃CH₂CH₂CH₂OH) (by GC, corrected for water) | ≥99.4 % |
ACS - Color (APHA) | ≤10 |
ACS - Residue after Evaporation | ≤0.0005 % |
ACS - Titrable Acid (µeq/g) | ≤0.8 |
ACS - Butyl Ether (C₈H₁₈O)(by GC) | ≤0.2 % |
Carbonyl Compounds - Butyraldehyde | ≤0.01 % |
Water (by KF, coulometric) | ≤0.05 % |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-300 nm | ≤0.01 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm | ≤0.02 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm | ≤0.04 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm | ≤0.10 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm | ≤0.50 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 215 nm | ≤1.00 |
Product Information (not specifications): | |
Boiling Point (typical)................. 117.7 °C. | |
Windows of Infrared Transmittance (0.1-mm path, 50-100% T), µm: | |
2.0 - 2.8 ; 4.3 - 6.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...