Chloroform ≥99.8% (by GC, corrected for water content) stabilised, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilization: ∼0.015% Stabilised with amylene
Management of Change (MOC) category = N

9174-03
JT9174-3CS 1522.92 USD
JT9174-3
Chloroform ≥99.8% (by GC, corrected for water content) stabilised, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Chloroform

For HPLC. UV cutoff 245nm. Stabilized with ~0.015% amylene. Lot analysis on label. Packaged under nitrogen.

Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7
Melting Pt: -63
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-280 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm ≤0.15
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤245
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.2 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
Assay (by GC, corrected for water, exclusive of preservative) ≥99.8 %
Chloride (Cl) ≤10 ppm
Preservative (amylene)(by wt) 0.01 - 0.02 %
Residue after Evaporation ≤2.0 ppm
Titrable Acid (µeq/g) ≤0.5
Water (by KF, coulometric) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR