Aniline ≥99% (by GC, corrected for water content), BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Aminobenzene, Phenylamine

Management of Change (MOC) category = N

9110-01 9110-03
JT9110-1CS 2211.81 USD
JT9110-1 JT9110-3
Aniline ≥99% (by GC, corrected for water content), BAKER ANALYZED® ACS
Aniline

Lot analysis on label.

Formula: C₆H₅NH₂
MW: 93.13 g/mol
Boiling Pt: 184
Melting Pt: -6.2
Density: 1.019 g/cm³ (20 °C)
Flash Pt: 76
Storage Temperature: Ambient
MDL Number: MFCD00007629
CAS Number: 62-53-3
EINECS: 200-539-3
UN: 1547
ADR: 6.1 II
Merck Index: 13,00661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fc5b5071f5a578057c8a7c1c4855ca964c4d840&vid=4f9c118a47c84e5b067d971f00a4eee6dc846d80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
NOTE: This product darkens to a reddish-brown colo r on storage.
Assay (C₆H₅NH₂) (by GC, corrected for water) ≥99.0 %
Color (APHA) (at time of packaging) ≤250
Residue after Ignition ≤0.005 %
Chlorobenzene (C₆H₅Cl) ≤0.01 %
Hydrocarbons Passes Test
Nitrobenzene (C₆H₅NO₂) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR