2-Propanol ≥99.7%, BAKER ANALYZED® HPLC for HPLC, for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

9095-02 9095-33 9095-03 9095-3H 9095-6K
JT9095-2CS 711.91 USD
JT9095-2 JT9095-33 TX9095-3HSEP JT9095-3 JT9095-3H JT9095-6K JT9095-88
2-Propanol ≥99.7%, BAKER ANALYZED® HPLC for HPLC, for spectrophotometry, J.T.Baker®
2-Propanol

For high performance liquid chromatography. UV cutoff at 205nm. Lot analysis on label.

Formula: (CH₃)₂CHOH
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm ≤0.020
Ultraviolet Absorbance (1.00-cm cell vs. water) - 225 nm ≤0.16
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤205
Assay (CH₃CHOHCH₃) ≥99.7 %
Residue after Evaporation ≤2.0 ppm
Water (by KF, coulometric) ≤0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR