Amyl acetate mixture of isomers, purified, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
n-Pentyl acetate, Acetic acid amyl ester, Acetic acid pentyl ester, Pentyl acetate
Management of Change (MOC) category = N
9094-03
9094-01
JT9094-3CS
2093.56
USD
JT9094-3
JT9094-1
Amyl acetate mixture of isomers, purified, J.T.Baker®
Amyl acetate
Formula:
C₇H₁₄O₂ MW: 130.19 g/mol Boiling Pt: 149 Melting Pt: -71 Density: 0.876 g/cm³ (20 °C) Flash Pt: 23 (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00009500 CAS Number: 628-63-7 EINECS: 211-047-3 UN: 1104 ADR: 3 III |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Identification (by IR) | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...