2-Propanol ≥99.5%, CMOS for microelectronic, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol

Management of Change (MOC) category = R

9079-05 9079-03
JT9079-5CS 1422.5 USD
JT9079-5 JT9079-3
2-Propanol ≥99.5%, CMOS for microelectronic, J.T.Baker®
2-Propanol
Formula: (CH₃)₂CHOH
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Microelectronic Use
Assay (CH₃CHOHCH₃) ≥ 99.5 %
Color (APHA) ≤ 10
Residue after Evaporation ≤ 4 ppm
Solubility in H₂O Passes Test
Water (H₂O)(by Karl Fischer titrn) ≤ 0.05 %
Acidity (µeq/g) ≤ 0.2
Alkalinity (µeq/g) ≤ 0.1
Chloride (Cl) ≤ 0.2 ppm
Phosphate (PO₄) ≤ 0.3 ppm
Trace Impurities - Aluminum (Al) ≤ 50.0 ppb
Arsenic and Antimony (as As) ≤ 10.0 ppb
Trace Impurities - Barium (Ba) ≤ 20.0 ppb
Trace Impurities - Beryllium (Be) ≤ 100.0 ppb
Trace Impurities - Bismuth (Bi) ≤ 100.0 ppb
Trace Impurities - Boron (B) ≤ 10.0 ppb
Trace Impurities - Cadmium (Cd) ≤ 20.0 ppb
Trace Impurities - Calcium (Ca) ≤ 50.0 ppb
Trace Impurities - Chromium (Cr) ≤ 20.0 ppb
Trace Impurities - Cobalt (Co) ≤ 20 ppb
Trace Impurities - Copper (Cu) ≤ 10.0 ppb
Trace Impurities - Gallium (Ga) ≤ 50 ppb
Trace Impurities - Germanium (Ge) ≤ 50.0 ppb
Trace Impurities - Gold (Au) ≤ 20 ppb
Heavy Metals (as Pb) ≤ 200.0 ppb
Trace Impurities - Iron (Fe) ≤ 50.0 ppb
Trace Impurities - Lead (Pb) ≤ 20.0 ppb
Trace Impurities - Lithium (Li) ≤ 50.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 20 ppb
Trace Impurities - Manganese (Mn) ≤ 15.0 ppb
Trace Impurities - Molybdenum (Mo) ≤ 100.0 ppb
Trace Impurities - Nickel (Ni) ≤ 10.0 ppb
Trace Impurities - Niobium (Nb) ≤ 100.0 ppb
Trace Impurities - Potassium (K) ≤ 100 ppb
Trace Impurities - Silicon (Si) ≤ 50 ppb
Trace Impurities - Silver (Ag) ≤ 20.0 ppb
Trace Impurities - Sodium (Na) ≤ 100.0 ppb
Trace Impurities - Strontium (Sr) ≤ 20.0 ppb
Trace Impurities - Tantalum (Ta) ≤ 100.0 ppb
Trace Impurities - Thallium (Tl) ≤ 10.0 ppb
Trace Impurities - Tin (Sn) ≤ 100 ppb
Trace Impurities - Titanium (Ti) ≤ 20 ppb
Trace Impurities - Vanadium (V) ≤ 100.0 ppb
Trace Impurities - Zinc (Zn) ≤ 50 ppb
Trace Impurities - Zirconium (Zr) ≤ 100.0 ppb
Particle Count - 0.2 µm and greater ≤ 5000 par/ml
Particle Count - 0.3 µm and greater ≤ 5000 par/ml
Particle Count - 0.5 µm and greater ≤ 50 par/ml
Particle Count - 1.0 µm and greater ≤ 8 par/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR