Methanol ≥99.9%, ULTRA RESI-ANALYZED® for gas chromatography, for purge and trap GC analysis, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

9077-02
JT9077-2CS 2266.53 USD
JT9077-2
Methanol ≥99.9%, ULTRA RESI-ANALYZED® for gas chromatography, for purge and trap GC analysis, J.T.Baker®
Methanol

BAKER ULTRA RESI-ANALYZED® Reagent—ultra high purity solvents and salts developed for organic residue extraction/concentration procedures. These solvents are fully characterized and lot controlled by ECD, FID, or other method-specific detectors. Advanced stabilizer systems are incorporated to ensure solvent stability and superior performance Solvents are suitable for exacting EPA sample cleanup protocols.

Formula: CH₃OH
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CH₃OH) (by GC, corrected for water) ≥99.9 %
Residue after Evaporation ≤1.0 ppm
Titrable Acid (µeq/g) ≤0.3
Titrable Base (µeq/g) ≤0.10
Water (by KF, coulometric) ≤0.08 %
Volatile Organic Trace Analysis - Below EPA 8260B CRQL Conforms / Fails
Performance Tested for Use in EPA Methods
500 Series for Drinking Water
600 Series for Wastewater
846 for Solid Waste


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR