Methanol ≥99.8% (by GC), BAKER ANALYZED® A.C.S. Reagent ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Wood alcohol, Methyl alcohol, Methyl hydroxide, Hydroxy methane, Carbinol, Methylol

Management of Change (MOC) category = N

9070-33 9070-05 9070-R 9070-03 9070-07 BAKR9070-01
JT9070-33CS 930.27 USD
JT9070-33 JT9070-5 JT9070-R JT9070-3 JT9070-7 JT9070-01
Methanol ≥99.8% (by GC), BAKER ANALYZED® A.C.S. Reagent ACS, J.T.Baker®
Methanol

The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.

Formula: CH₃OH
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CH₃OH) (by GC) ≥ 99.8 %
Substances Darkened by H₂SO₄ Passes Test
Substances Reducing KMnO₄ Passes Test
Solubility in H₂O Passes Test
Color (APHA) ≤ 10
Water (by KF, coulometric) ≤ 0.1 %
Residue after Evaporation ≤ 0.001 %
Carbonyl Compounds - Acetaldehyde ≤ 0.001 %
Carbonyl Compounds - Acetone ≤ 0.001 %
Carbonyl Compounds - Formaldehyde ≤ 0.001 %
Titrable Acid (µeq/g) ≤ 0.3
Titrable Base (µeq/g) ≤ 0.2
Heavy Metals (as Pb) ≤ 0.5 ppm
Trace Impurities - Copper (Cu) ≤ 0.10 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Trace Impurities - Nickel (Ni) ≤ 0.1 ppm
Date Received: __ __ /__ __ /__ __
Date Opened: __ __ /__ __ /__ __
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR