1-Butanol ≥99.4% (by GC, corrected for water content), BAKER ANALYZED® A.C.S. Reagent ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

9054-01 9054-03 9054-07 9054-R
JT9054-1CS 1212.38 USD
JT9054-1 JT9054-3 JT9054-7 JT9054-R
1-Butanol ≥99.4% (by GC, corrected for water content), BAKER ANALYZED® A.C.S. Reagent ACS
1-Butanol

The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.

Formula: H₃C(CH₂)₂CH₂OH
Boiling Pt: 117.6 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.812 g/cm³ (20 °C)
Flash Pt: 37 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3,III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CH₃CH₂CH₂CH₂OH) (by GC, corrected for water) ≥99.4 %
Color (APHA) ≤10
Residue after Evaporation ≤0.0050 %
Titrable Acid (meq/g) ≤0.0008
Butyl Ether (C₈H₁₈O)(by GC) ≤0.2 %
Carbonyl Compounds - Butyraldehyde ≤0.01 %
Water (by KF, coulometric) ≤0.1 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR