Benzyl alcohol ≥99.5%, BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = N
9050-01
9050-03
JT9050-1CS
2132.42
USD
JT9050-1
JT9050-03
Benzyl alcohol ≥99.5%, BAKER ANALYZED®, J.T.Baker®
Benzyl alcohol
Lot analysis on label.
Formula:
C₆H₅CH₂OH Boiling Pt: 205 °C (1013 hPa) Melting Pt: –15.3 °C Density: 1.05 g/cm³ (20 °C) Flash Pt: 101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004599 CAS Number: 100-51-6 EINECS: 202-859-9 Merck Index: 13,01126 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (C₆H₅CH₂OH) (by GC, corrected for water) | ≥99.5 % |
Color (APHA) | ≤10 |
Residue after Ignition | ≤0.005 % |
Solubility in H₂O | Passes Test |
Chlorinated Compounds (as Cl) | Passes Test |
Water (H₂O)(by Karl Fischer titrn) | ≤0.05 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...