Benzyl alcohol 98.0-100.5% NF, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = R
9040-03
JT9040-03CS
2441.3
USD
JT9040-03
Benzyl alcohol 98.0-100.5% NF, J.T.Baker®
Benzyl alcohol
Caution: This product is not intended for use in preparing parenteral dosage forms.
Formula:
C₆H₅CH₂OH Boiling Pt: 205 °C (1013 hPa) Melting Pt: –15.4…–15.4 °C Density: 1.05 g/cm³ (20 °C) Flash Pt: 100.40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004599 CAS Number: 100-51-6 Merck Index: 13,01126 |
|
Specification Test Results
GMP Manufactured Product | |
Meets N.F. Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
NF - Assay | 98.0 - 100.5 % |
NF - Clarity of Solution | Passes Test |
NF - Color of Solution | Passes Test |
NF - Identification | Passes Test |
NF - Peroxide Value | ≤5 |
NF - Refractive Index, n²ºD | 1.538 - 1.541 |
NF - Benzaldehyde and Other Related Substances - Benzaldehyde | ≤0.15 % |
NF - Related Substances - Cyclohexylmethanol | ≤0.10 % |
of Benzyl Alcohol | ≤0.04 % |
NF - Benzaldehyde and Other Related Substances - Total Other Peaks | |
NF - Benzaldehyde and Other Related Substances - Total Other Peaks | ≤0.3 % |
Greater Than RT of Benzyl Alcohol | ≤0.3 % |
NF - Acidity | Passes Test |
NF - Residue on Evaporation | ≤0.05 % |
NF - Benzene | ≤2 ppm |
This product is not intended for use in preparing parenteral dosage | |
forms. | |
The Class 1 solvent Benzene is likely to be present below 2 ppm. The | |
actual analysis of Benzene is reported for each lot. | |
The Class 2 solvent Toluene is likely to be present and is process | |
controlled to <30 ppm. | |
No other Class 1, 2 or 3 residual solvents are likely to be present. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...