Isoamyl alcohol ≥98.5%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

9038-03 9038-01
JT9038-3CS 4234.66 USD
JT9038-3 JT9038-1
Isoamyl alcohol ≥98.5%, BAKER ANALYZED® ACS, J.T.Baker®
Isoamyl alcohol

Lot analysis on label.

Formula: (CH₃)₂CHCH₂CH₂OH
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –117 °C
Density: 0.812 g/cm³ (20 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00002934
CAS Number: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3,III
Merck Index: 13,05214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=170c60e26c7b4edab6e0686f91d04e676da16e6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay ((CH₃)₂CHCH₂CH₂OH) (by GC, corrected for water) ≥98.5 %
Residue after Evaporation ≤0.003 %
Titrable Acid (meq/g) ≤0.002
Acids and Esters (as amyl acetate) ≤0.2 %
Carbonyl Compounds (as HCHO) (by polarography) ≤0.1 %
Water (H₂O)(by Karl Fischer titrn) ≤0.5 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR