Acetonitrile ≥99.9% (by GC) for HPLC, for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

9012-03
JT9012-3CS 2398.58 USD
JT9012-3
Acetonitrile ≥99.9% (by GC) for HPLC, for spectrophotometry, J.T.Baker®
Acetonitrile

BAKER ANALYZED™ HPLC Reagent—High purity solvents for use in analytical and preparative separations. All are optimized for all of your Liquid Chromatography applications, including HPLC, UHPLC, gel permeation chromatography and UV-spectrophotometric analysis.

Formula: H₃CCN
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay (GC) ≥99.9 %
Color (APHA) ≤10
Water (H₂O) ≤0.02 %
Gradient Elution Test (a.u.) - 254 nm ≤0.005
Residue after Evaporation ≤3 ppm
Titrable Acid (µeq/g) ≤8
Titrable Base (µeq/g) ≤0.6
Ultraviolet Absorbance (1.00 cm) - 200 nm ≤0.1
Ultraviolet Absorbance (1.00 cm) - 210 nm ≤0.05
Ultraviolet Absorbance (1.00 cm) - 220 nm ≤0.03
Ultraviolet Absorbance (1.00 cm) - 254 nm ≤0.01
Ultraviolet Absorbance (1.00 cm) - 280 nm ≤0.01
Ultraviolet Absorbance (1.00 cm) - 350 nm ≤0.01
Ultraviolet Absorbance (1.00 cm) - 400 nm ≤0.01
Ultraviolet Absorbance (1.00 cm) - UV Cut-off, nm ≤190
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR