Acetone ≥99.5%, BAKER ANALYZED®, Electronic Grade, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = R

9009-07
JT9009-7EA 231.83 USD
JT9009-7
Acetone ≥99.5%, BAKER ANALYZED®, Electronic Grade, J.T.Baker®
Acetone
Formula: CH₃COCH₃
Boiling Pt: 56.2 °C (1013 hPa)
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Microelectronic Use
Assay ((CH₃)₂CO) (by GC, corrected for water) ≥99.5 %
Color (APHA) ≤10
Residue after Evaporation ≤0.001 %
Solubility in H₂O Passes Test
Titrable Acid (meq/g) ≤0.0003
Titrable Base (meq/g) ≤0.0006
Aldehydes (as HCHO) ≤0.002 %
Isopropyl Alcohol (CH₃CHOHCH₃)(by GC) ≤0.05 %
Methanol (CH₃OH)(by GC) ≤0.05 %
Heavy Metals (as Pb) ≤0.20 ppm
Substances Reducing Permanganate Passes Test
Water (H₂O) ≤0.5 %
Chloride (Cl) ≤0.2 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤0.050 ppm
Nickel (Ni) ≤0.1 ppm
Manganese (Mn) ≤0.100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR