Acetone ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® A.C.S. Reagent ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

9006-05 9006-R 9006-22 9006-07 9006-33 9006-03 BAKR9006-01 BAKR9006-06
JT9006-5CS 1229.24 USD
JT9006-5 JT9006-R JT9006-1 JT9006-22 JT9006-7 JT9006-33 JT9006-3 JT9006-ZT JT9006-01 JT9006-06
Acetone ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® A.C.S. Reagent ACS, J.T.Baker®
Acetone

The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.


  • Produced using proven distillation methods and rigid testing protocols
  • Statistical Process Control used in purification and packaging to ensure lot-to-lot consistency
  • Statistical Quality Control and suitability-for-use tests to ensure product performance

Formula: CH₃COCH₃
Boiling Pt: 56.2 °C (1013 hPa)
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay ((CH₃)₂CO) (by GC, corrected for water) ≥99.5 %
Color (APHA) ≤10
Residue after Evaporation ≤0.001 %
Solubility in H₂O Passes Test
Titrable Acid (meq/g) ≤0.0003
Titrable Base (meq/g) ≤0.0006
Aldehydes (as HCHO) ≤0.002 %
Isopropyl Alcohol (CH₃CHOHCH₃)(by GC) ≤0.05 %
Methanol (CH₃OH)(by GC) ≤0.05 %
Heavy Metals (as Pb) ≤1 ppm
Substances Reducing Permanganate Passes Test
Water (H₂O) ≤0.5 %
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤0.1 ppm
Nickel (Ni) ≤0.1 ppm
Date Received: __ __ /__ __ /__ __
Date Opened: __ __ /__ __ /__ __
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR