EDTA (ethylenediamine tetraacetic acid) 99.4-100.6%, powder, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, Edetic Acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, EDTA, (Ethylenedinitrilo)tetraacetic acid, Idranal® II

Management of Change (MOC) category = N

8991-01
JT8991-1CS 1185.48 USD
JT8991-1
EDTA (ethylenediamine tetraacetic acid) 99.4-100.6%, powder, BAKER ANALYZED® ACS, J.T.Baker®
EDTA (Ethylenediamine tetraacetic acid)

The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.

Formula: C₁₀H₁₆N₂O₈
MW: 292.25 g/mol
Boiling Pt: 614 °C (1013 hPa)
Melting Pt: 220 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 13,03546
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (C₁₀H₁₆N₂O₈) 99.4 - 100.6 %
ACS - Insoluble in Dilute NH₄OH ≤0.005 %
ACS - Residue after Ignition ≤0.2 %
ACS - Nitrilotriacetic Acid ≤0.1 %
ACS - Calcium (Ca) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Iron (Fe) ≤0.005 %
Trace Impurities - Magnesium (Mg) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR