J.T.Baker® BAKERBOND® Protein Precipitation Plates
Supplier: Avantor
J.T.Baker® BAKERBOND® protein precipitation plates enable fast protein removal from biological samples.
- Protein removal from plasma
- Ideal prior to LC-MS applications
- 96 well plate format for multiple samples
- Vacuum manifold can be used (8183-VM)
- Collection plates available
Proteins crash out of solution when the standard protocol is followed, this process removes proteins from the matrix that could interfere with LC-MS analyses and also avoids any protein precipitation during the HPLC analyses.