J.T.Baker® BAKERBOND Speedisk®, Oil and Grease Standards Kit
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
BAKR8030-00
JT8030-0PK 185.07 USD
JT8030-0 JT80300
J.T.Baker® BAKERBOND Speedisk®, Oil and Grease Standards Kit
Standards Environmental Analysis Standards
51 ml/bottle mixture of 2 mg/mLl Stearic Acid and 2 mg/ml n-Hexadecane in ULTRA RESIANALYZED Acetone.
Packaging: Bottles have a rubber stopper and are foil sealed.