Potassium dihydrogen phosphate 98.0-100.5% (dried basis) NF, ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Monopotassiumphosphate, Potassium orthophosphate monobasic, prim.-Potassium phosphate, Potassium dihydrogenorthophosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

Management of Change (MOC) category = R

7390-19
JT7390-19EA 1662.11 USD
JT7390-19
Potassium dihydrogen phosphate 98.0-100.5% (dried basis) NF, ACS, J.T.Baker®
Potassium dihydrogen phosphate
Formula: KH₂PO₄
MW: 136.09 g/mol
MDL Number: MFCD00011401
CAS Number: 7778-77-0

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay (KH₂PO₄) (dried basis) 98.0 - 100.5 %
NF - Arsenic (As) ≤3 ppm
NF - Fluoride (F) ≤10 mg/kg
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Insoluble Substances ≤0.2 %
NF - Lead (Pb) ≤5 ppm
NF - Loss on Drying at 105°C ≤1.0 %
ACS - Assay (KH₂PO₄) (by acidimetry) ≥99.0 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Insoluble Matter ≤0.01 %
ACS - Iron (Fe) ≤0.002 %
ACS - Loss on Drying at 105°C ≤0.2 %
ACS - Nitrogen Compounds (as N) ≤0.001 %
ACS - pH of 5% Solution at 25°C 4.1 - 4.5
ACS - Sodium (Na) ≤0.005 %
ACS - Sulfate (SO₄) ≤0.003 %
Solubility (10 in 100) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR