J.T.Baker® BAKERBOND®, Hydrophobic Interaction Chromatography Media
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
7285-00
JT7285-0EA
42507.6
USD
JT7285-0
J.T.Baker® BAKERBOND®, Hydrophobic Interaction Chromatography Media
Protein Purification
Protein Purification Resins
Silica based media used for the purification of proteins based on differential salt induced hydrophobicity.
Mixed mode provides unique selectivity with its hydrophobic interaction and ion exchange functionalities.