Acetic acid glacial_AcOH 99,0, ULTREX® II, ultrapure, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

M.O.C. Category N

6903-05
JT6903-5EA 909.38 USD
JT6903-5
Acetic acid glacial_AcOH 99,0, ULTREX® II, ultrapure, J.T.Baker®
Acetic acid

For trace metal analysis. Certificate of lot analysis provided.

Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
IMPORTANT: Material will freeze if stored below 17 °C (63°F).
Assay (CH₃COOH) ≥99.0 %
Trace Impurities - Aluminum (Al) ≤50 ppt
Trace Impurities - Antimony (Sb) ≤50 ppt
Trace Impurities - Arsenic (As) ≤50 ppt
Trace Impurities - Barium (Ba) ≤10 ppt
Trace Impurities - Beryllium (Be) ≤10 ppt
Trace Impurities - Bismuth (Bi) ≤10 ppt
Trace Impurities - Cadmium (Cd) ≤10 ppt
Trace Impurities - Calcium (Ca) ≤50 ppt
Trace Impurities - Cerium (Ce) ≤10 ppt
Trace Impurities - Cesium (Cs) ≤10 ppt
Trace Impurities - Chromium (Cr) ≤10 ppt
Trace Impurities - Cobalt (Co) ≤10 ppt
Trace Impurities - Copper (Cu) ≤10 ppt
Trace Impurities - Dysprosium (Dy) ≤1 ppt
Trace Impurities - Erbium (Er) ≤1 ppt
Trace Impurities - Europium (Eu) ≤1 ppt
Trace Impurities - Gadolinium (Gd) ≤1 ppt
Trace Impurities - Gallium (Ga) ≤10 ppt
Trace Impurities - Germanium (Ge) ≤10 ppt
Trace Impurities - Hafnium (Hf) ≤10 ppt
Trace Impurities - Holmium (Ho) ≤1 ppt
Trace Impurities - Indium (In) ≤1 ppt
Trace Impurities - Iron (Fe) ≤50 ppt
Trace Impurities - Lanthanum (La) ≤1 ppt
Trace Impurities - Lead (Pb) ≤10 ppt
Trace Impurities - Lithium (Li) ≤10 ppt
Trace Impurities - Lutetium (Lu) ≤10 ppt
Trace Impurities - Magnesium (Mg) ≤50 ppt
Trace Impurities - Manganese (Mn) ≤10 ppt
Trace Impurities - Molybdenum (Mo) ≤10 ppt
Trace Impurities - Neodymium (Nd) ≤1 ppt
Trace Impurities - Nickel (Ni) ≤50 ppt
Trace Impurities - Platinum (Pt) ≤50 ppt
Trace Impurities - Potassium (K) ≤50 ppt
Trace Impurities - Praseodymium (Pr) ≤1 ppt
Trace Impurities - Rhenium (Re) ≤10 ppt
Trace Impurities - Rhodium (Rh) ≤50 ppt
Trace Impurities - Rubidium (Rb) ≤10 ppt
Trace Impurities - Ruthenium (Ru) ≤50 ppt
Trace Impurities - Samarium (Sm) ≤1 ppt
Trace Impurities - Scandium (Sc) ≤10 ppt
Trace Impurities - Selenium (Se), For Information Only
Trace Impurities - Silver (Ag) ≤50 ppt
Trace Impurities - Sodium (Na) ≤100 ppt
Trace Impurities - Strontium (Sr) ≤10 ppt
Trace Impurities - Tellurium (Te) ≤1 ppt
Trace Impurities - Terbium (Tb) ≤1 ppt
Trace Impurities - Thallium (Tl) ≤10 ppt
Trace Impurities - Thorium (Th) ≤1 ppt
Trace Impurities - Thulium (Tm) ≤1 ppt
Trace Impurities - Tin (Sn) ≤50 ppt
Trace Impurities - Titanium (Ti) ≤10 ppt
Trace Impurities - Tungsten (W) ≤10 ppt
Trace Impurities - Uranium (U) ≤1 ppt
Trace Impurities - Vanadium (V) ≤10 ppt
Trace Impurities - Ytterbium (Yb) ≤1 ppt
Trace Impurities - Yttrium (Y) ≤1 ppt
Trace Impurities - Zinc (Zn) ≤50 ppt
Trace Impurities - Zirconium (Zr) ≤10 ppt
Certificate Provided Reports Actual Lot Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR