N-Methyl-2-pyrrolidone (NMP) ≥99.8%, Finyte® for microelectronic, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, N-Methylpyrrolidinone, N-Methyl-2-pyrrolidinone, N-Methyl-alpha-pyrrolidone, M-Pyrol, N-methylpyrrolidone, N-Methyl-gamma-butyrolactam, 1-Methyl-2-pyrrolidone, NMP, N-Methylpyrrolidone, N-Methyl-2-pyrrolidone, Methyl Pyrrolidone

Management of Change (MOC) category = R

6337-05
JT6337-05CS 681.86 USD
JT6337-05
N-Methyl-2-pyrrolidone (NMP) ≥99.8%, Finyte® for microelectronic, J.T.Baker®
N-Methyl-2-pyrrolidone (NMP)
Formula: C₅H₉NO
MW: 99.13 g/mol
Density: 1.03 g/cm³
Flash Pt: 96.00 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Microelectronic Use
Assay (C₅H₉NO) ≥99.8 %
Color (APHA) ≤30
Water (H₂O)(by Karl Fischer titrn) ≤300 ppm
Free Amines (as CH₃NH₂) ≤50 ppm
Chloride (Cl) ≤0.5 ppm
Phosphate (PO₄) ≤1 ppm
Trace Impurities - Aluminum (Al) ≤10.0 ppb
Arsenic and Antimony (as As) ≤10.0 ppb
Trace Impurities - Barium (Ba) ≤10.0 ppb
Trace Impurities - Beryllium (Be) ≤10.0 ppb
Trace Impurities - Bismuth (Bi) ≤10.0 ppb
Trace Impurities - Boron (B) ≤10.0 ppb
Trace Impurities - Cadmium (Cd) ≤10.0 ppb
Trace Impurities - Calcium (Ca) ≤10.0 ppb
Trace Impurities - Chromium (Cr) ≤10.0 ppb
Trace Impurities - Cobalt (Co) ≤10.0 ppb
Trace Impurities - Copper (Cu) ≤10.0 ppb
Trace Impurities - Gallium (Ga) ≤10.0 ppb
Trace Impurities - Germanium (Ge) ≤10.0 ppb
Trace Impurities - Gold (Au) ≤10.0 ppb
Trace Impurities - Iron (Fe) ≤10.0 ppb
Trace Impurities - Lead (Pb) ≤10.0 ppb
Trace Impurities - Lithium (Li) ≤10.0 ppb
Trace Impurities - Magnesium (Mg) ≤10.0 ppb
Trace Impurities - Manganese (Mn) ≤10.0 ppb
Trace Impurities - Molybdenum (Mo) ≤10.0 ppb
Trace Impurities - Nickel (Ni) ≤10.0 ppb
Trace Impurities - Niobium (Nb) ≤10.0 ppb
Trace Impurities - Potassium (K) ≤10.0 ppb
Trace Impurities - Silver (Ag) ≤10.0 ppb
Trace Impurities - Sodium (Na) ≤10.0 ppb
Trace Impurities - Strontium (Sr) ≤10.0 ppb
Trace Impurities - Tantalum (Ta) ≤10.0 ppb
Trace Impurities - Thallium (Tl) ≤10.0 ppb
Trace Impurities - Tin (Sn) ≤10.0 ppb
Trace Impurities - Titanium (Ti) ≤10.0 ppb
Trace Impurities - Vanadium (V) ≤10.0 ppb
Trace Impurities - Zinc (Zn) ≤10.0 ppb
Trace Impurities - Zirconium (Zr) ≤10.0 ppb
Particle Count - 0.3 µm and greater (Rion KS42AF)
Particle Count - 0.5 µm and greater (Rion KS42AF) ≤100 par/ml
Particle Count - 1.0 µm and greater (Rion KS42AF) ≤10 par/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR