J.T.Baker® HYDRA-POINT™, Coulometric Reagents for Water Determination by Karl Fischer Titration
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
6285-01 6287-02
JT6280-1CS 1401.42 USD
JT6280-1 JT6280-2 JT6281-0 JT6283-1 JT6284-1 JT6284-2 JT6285-1 JT6285-2 JT6287-1 JT6287-2
J.T.Baker® HYDRA-POINT™, Coulometric Reagents for Water Determination by Karl Fischer Titration
Karl Fischer Titrants and Reagents
HYDRA-POINT® coulometric reagents are designed to deliver faster analyses, lower detection limits and increased accuracy.
  • Pyridine- and chlorocarbon-free vessel solution
  • The fritless/diaphragmless reagent works both in cells without and with diaphragm
  • High water capacity, where the anolyte can take over 1000 mg of water per charge
These products feature lower background noise that gives a better signal/noise ratio, resulting in a significant increase in the lower detection limit—to as low as 1 ppm.