Phenolphthalein alcoholic solution, BAKER ANALYZED® USP Test Solution (TS), J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

Management of Change (MOC) category = N

5927-01
JT5927-1CS 1170.43 USD
JT5927-1
Phenolphthalein alcoholic solution, BAKER ANALYZED® USP Test Solution (TS), J.T.Baker®
Phenolphthalein

Certifications: Meets test solution (T.S.) specifications for testing USP/NF monographs.

Formula: C₂₀H₁₄O₄
Flash Pt: 13 °C
MDL Number: MFCD00005913
CAS Number: 77-09-8
UN: 1170
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Visual Transition Interval - pH…8.0 (Colorless) Passes Test
Visual Transition Interval - pH…10.0 (Pink) Passes Test
Meets test solution (T.S.) specifications for testing USP/NF monographs.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR