Barium chloride T.S., BAKER ANALYZED® PharmaTest™, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

5907-01
JT5907-1CS 584.52 USD
JT5907-1
Barium chloride T.S., BAKER ANALYZED® PharmaTest™, J.T.Baker®
Barium chloride

Certifications: Meets test solution (T.S.) specifications for testing USP/NF monographs.

Formula: BaCl₂
MW: 208.24 g/mol
MDL Number: MFCD00003445
CAS Number: 10361-37-2
UN: 1564
ADR: 6.1,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (BaCl₂·2H₂O), w/v 11.6 - 12.4 %
Meets test solution (T.S.) specifications for testing USP/NF monographs.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR