Sodium hydroxide 6 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
Management of Change (MOC) category = NI
5672-02
BAKR5672-03
JT5672-3CS
602.58
USD
JT5672-3
JT5672-2
JT5672-03
Sodium hydroxide 6 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
Sodium hydroxide
J.T.Baker® volumetric and analytical solutions. These solutions are suitable for use in ACS, USP and NF compendial methods and general laboratory applications.
J.T.Baker® volumetric and analytical solutions. These solutions are suitable for use in ACS, USP and NF compendial methods and general laboratory applications.
Formula:
NaOH Boiling Pt: 109 °C (1013 hPa) Density: ≤1.11 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Protect from air to avoid absorption of carbon dioxide. | |
Normality | 5.98 - 6.02 |
Chloride (Cl) | ≤5 ppm |
ACS - Heavy Metals (as Pb) | ≤1 ppm |
Trace Impurities - Iron (Fe) | ≤0.5 ppm |
Standardization at 25°C traceable to NIST Standard Reference Material. | |
SRM No… |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...