Potassium hydroxide 0.1 N in ethanol, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

Management of Change (MOC) category = N

5645-02
JT5645-2CS 836.88 USD
JT5645-2
Potassium hydroxide 0.1 N in ethanol, BAKER ANALYZED® volumetric solution, J.T.Baker®
Potassium hydroxide

Standardized at 25°C against chemicals whose certification is traceable to NIST. Lot standardization value, accurate to +/- 1 part per 1000, provided on label.

Formula: KOH
MW: 56.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.0995 - 0.1005
Appearance Passes Test
Standardization at 25°C traceable to NIST Standard Reference Material. Theactual lot standardization value, accurate to within +/- 1 part per 1000, isprovided on the label.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR