Potassium hydroxide 0.5 N in ethanol, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

Management of Change (MOC) category = N

5644-02
JT5644-2CS 914.69 USD
JT5644-2
Potassium hydroxide 0.5 N in ethanol, BAKER ANALYZED® volumetric solution, J.T.Baker®
Potassium hydroxide

J.T.Baker® volumetric and analytical solutions. These solutions are suitable for use in ACS, USP and NF compendial methods and general laboratory applications. 

Formula: KOH
MW: 56.11 g/mol
Flash Pt: 13.00 °C
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.495 - 0.505
Appearance Passes Test
Standardization at 25°C traceable to NIST Standard Reference Material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR