Sodium hydroxide 0.25 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

Management of Change (MOC) category = N

5638-03 5638-02
JT5638-3CS 537.14 USD
JT5638-3 JT5638-2
Sodium hydroxide 0.25 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Sodium hydroxide

Standardization at 25 °C traceable to NIST Standard Reference Material.

Formula: NaOH
Boiling Pt: ∼105 °C (1013 hPa)
Density: 1.02 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Protect from air to avoid absorption of carbon dioxide.
Normality 0.2490 - 0.2510
Chloride (Cl) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR