Sodium hydroxide 2 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
Management of Change (MOC) category = NI
5633-02
5633-03
JT5633-2CS
515.8
USD
JT5633-2
JT5633-3
Sodium hydroxide 2 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Sodium hydroxide
Standardization at 25 °C traceable to NIST Standard Reference Material.
Formula:
NaOH Boiling Pt: 109 °C (1013 hPa) Density: ≤1.11 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Protect from air to avoid absorption of carbon dioxide. | |
Normality | 1.990 - 2.010 |
Chloride (Cl) | ≤5 ppm |
ACS - Heavy Metals (as Pb) | ≤1 ppm |
Trace Impurities - Iron (Fe) | ≤0.5 ppm |
Standardization at 25°C traceable to NIST Standard Reference Material. | |
SRM No… |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...