EDTA disodium salt dihydrate 0.1 M, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate

Management of Change (MOC) category = NI

5632-02
JT5632-2CS 826.44 USD
JT5632-2
EDTA disodium salt dihydrate 0.1 M, BAKER ANALYZED® volumetric solution, J.T.Baker®
EDTA disodium salt dihydrate

Standardization at 25 °C traceable to NIST Standard Reference Material.

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Density: 1.02 g/cm³
MDL Number: MFCD00150037
CAS Number: 6381-92-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Molarity (Mol/L) 0.098 - 0.102
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR